Invitrogen
Manufacturer Code:PA575348
Catalog # PIPA575348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 711-760 of Human BAI1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor B1; BAI1 Brain-specific angiogenesis inhibitor 1; Brain-specific angiogenesis inhibitor 1; Vasculostatin-120; Vasculostatin-40; Vstat-40; Vstat120; Vstat40
Gene Aliases: ADGRB1; B830018M07Rik; BAI1; GDAIF; mKIAA4089; R75078
UniProt ID: (Human) O14514, (Mouse) Q3UHD1, (Rat) C0HL12
Entrez Gene ID: (Human) 575, (Mouse) 107831, (Rat) 362931
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.