R&D Systems
Manufacturer Code:AF4969SP
Catalog # AF4969SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human BAI1 Ala31-Thr879 Accession # O14514 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor B1; Brain-specific angiogenesis inhibitor 1; brain-specific angiogenesis inhibitor 1 FLJ41988 GDAIF; Vasculostatin-120; Vasculostatin-40; Vstat120; Vstat40
Gene Aliases: ADGRB1; BAI1; GDAIF
UniProt ID: (Human) O14514
Entrez Gene ID: (Human) 575
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.