Novus Biologicals
Manufacturer Code:NBP191067
Catalog # NBP191067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SLGLLCAELRGGSGGEPAKKRSKLERSVYAGLQTASVEKAQCKKSSCQGGLAPSVAHRVAQLKPKVKSKGLPTGLSSFQQKEATPGGRIREKLSRAK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAH and coiled-coil domain-containing protein 1; BAH and coiled-coil domain-containing protein 1 BAH domain and coiled-coil containing 1 BAH domain-containing protein 2 BAHD2 bromo adjacent homology domain-containing protein 2 FLJ23058 KIAA1447; BAH domain-containing protein 2; Bromo adjacent homology domain-containing protein 2
Gene Aliases: BAHCC1; BAHD2; KIAA1447
UniProt ID: (Human) Q9P281
Entrez Gene ID: (Human) 57597
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.