Proteintech
Manufacturer Code:666611IG150UL
Catalog # 50-173-7079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B8D3 |
Immunogen | BAG6 Fusion Protein Ag24525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BAG 6 BAG6 BAT3 D6S52E G3 HLA B associated transcript 3 Protein G3; BAG family molecular chaperone regulator 6; BAG-6; BCL2-associated athanogene 6; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; Protein G3; Protein Scythe; scythe
Gene Aliases: 2410045D21Rik; AA408914; BAG-6; BAG6; BAT3; D17H6S52E; D6S52E; G3; Scythe
UniProt ID: (Human) P46379, (Rat) Q9WTN8, (Mouse) Q9Z1R2
Entrez Gene ID: (Human) 7917, (Rat) 94342, (Mouse) 224727
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.