Invitrogen
Manufacturer Code:PA562770
Catalog # PIPA562770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BAG6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 6; BAG-6; BAG-6 BAT3 D6S52E G3 BAG family molecular chaperone regulator 6 HLA-B associated transcript 3 HLA-B associated transcript-3 HLA-B-associated transcript 3 large proline-rich protein BAG6 large proline-rich protein BAT3 protein G3 protein Scythe scythe; BCL2-associated athanogene 6; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; Protein G3; Protein Scythe; scythe
Gene Aliases: BAG-6; BAG6; BAT3; D6S52E; G3
UniProt ID: (Human) P46379
Entrez Gene ID: (Human) 7917
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.