Abnova
Manufacturer Code:H00009529M02A
Catalog # 89-106-096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E3 |
Immunogen | BAG5 (AAH505511 a.a. &bsim 447 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant BAG5.
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 5; BAG-5; BAG-family molecular chaperone regulator-5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5
Gene Aliases: BAG-5; BAG5; KIAA0873
UniProt ID: (Human) Q9UL15
Entrez Gene ID: (Human) 9529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.