Novus Biologicals
Manufacturer Code:NB10056091
Catalog # NB10056091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 168-187 (KKQPSLPLSEDAHPSVAKIN) of human BAG-5 was used as immunogen GenBank no. gi 6631077 ref NP_004864.1 . The amino acid sequence used for immunogen is 100% consevered with chicken and dog BAG-5 95% conserved with mouse BAG-5 and 94% conserved with rat BAG-5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 5; BAG family molecular chaperone regulator 5 BAG-5 BAG-family molecular chaperone regulator-5 Bcl-2-associated athanogene 5 BCL2-associated athanogene 5 KIAA0873; BAG-5; BAG-family molecular chaperone regulator-5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5
Gene Aliases: BAG-5; BAG5; KIAA0873
UniProt ID: (Human) Q9UL15
Entrez Gene ID: (Human) 9529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.