Novus Biologicals
Manufacturer Code:NBP292791100UL
Catalog # NB027448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 198-447 of human BAG5 (NP_004864.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 5; BAG family molecular chaperone regulator 5 BAG-5 BAG-family molecular chaperone regulator-5 Bcl-2-associated athanogene 5 BCL2-associated athanogene 5 KIAA0873; BAG-5; BAG-family molecular chaperone regulator-5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5
Gene Aliases: BAG-5; BAG5; KIAA0873
UniProt ID: (Human) Q9UL15
Entrez Gene ID: (Human) 9529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.