Invitrogen
Manufacturer Code:PA519047
Catalog # PIPA519047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GNATSDHPNNQDQSS) corresponding to the internal amino acids of BAG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 4; BAG-4; BAG-family molecular chaperone regulator-4; BAG4 BCL2-associated athanogene 4 BAG-4 SODD BAG-family molecular chaperone regulator-4 silencer of death domains; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; Silencer of death domains
Gene Aliases: BAG-4; BAG4; SODD
UniProt ID: (Human) O95429
Entrez Gene ID: (Human) 9530
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.