Invitrogen
Manufacturer Code:PA5106815
Catalog # PIPA5106815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BAG4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410112I15Rik BAG cochaperone 4 BAG family molecular chaperone regulator 4 BAG4 BAG-4 BAG-family molecular chaperone regulator-4 BCL2 associated athanogene 4 Bcl-2-associated athanogene 4 BCL2-associated athanogene 4 Silencer of death domains Sodd; BAG family molecular chaperone regulator 4; BAG-4; BAG-family molecular chaperone regulator-4; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; Silencer of death domains
Gene Aliases: 2410112I15Rik; BAG-4; BAG4; SODD
UniProt ID: (Human) O95429, (Mouse) Q8CI61
Entrez Gene ID: (Human) 9530, (Mouse) 67384, (Rat) 361167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.