Invitrogen
Manufacturer Code:PA141498
Catalog # 01-675-323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues S(2) A L R R S G Y G P S D G P S(16) of human SODD/BAG4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 4; BAG-4; BAG-4 SODD; BAG-family molecular chaperone regulator-4; Bcl-2-associated athanogene 4; BCL2 associated athanogene 4; BCL2-associated athanogene 4; Silencer of death domains
Gene Aliases: 2410112I15Rik; BAG-4; BAG4; SODD
UniProt ID: (Human) O95429, (Mouse) Q8CI61
Entrez Gene ID: (Dog) 607954, (Human) 9530, (Bovine) 516353, (Mouse) 67384, (Rat) 361167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.