Invitrogen
Manufacturer Code:PA578854
Catalog # PIPA578854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Bag3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 3; BAG family molecular chaperone regulator 3 BAG-3 Bcl-2-associated athanogene 3 Bcl-2-binding protein Bis Docking protein CAIR-1 BAG3 BIS; BAG-3; Bcl-2-associated athanogene 3; Bcl-2-binding protein Bis; Bcl-2-interacting death suppressor; BCL2-associated athanogene 3; BCL2-binding athanogene 3; Docking protein CAIR-1
Gene Aliases: AA407278; BAG-3; BAG3; BIS; CAIR-1; MFM6; mg638; MNCb-2243
UniProt ID: (Human) O95817, (Mouse) Q9JLV1
Entrez Gene ID: (Human) 9531, (Mouse) 29810, (Rat) 293524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.