Novus Biologicals
Manufacturer Code:NBP259476100UL
Catalog # NBP259476X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 29000000000 |
Immunogen | Recombinant human BAG2 (1-211aa) purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 2; BAG family molecular chaperone regulator 2 BAG-2 BAG-family molecular chaperone regulator-2 Bcl-2-associated athanogene 2 BCL2-associated athanogene 2 dJ417I1.2 dJ417I1.2 (BAG-family molecular chaperone regulator 2) KIAA0576 MGC149462; BAG-2; BAG-family molecular chaperone regulator-2; Bcl-2-associated athanogene 2; BCL2-associated athanogene 2; dJ417I1.2 (BAG-family molecular chaperone regulator 2)
Gene Aliases: BAG-2; BAG2; dJ417I1.2
UniProt ID: (Human) O95816
Entrez Gene ID: (Human) 9532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.