Invitrogen
Manufacturer Code:PA527701
Catalog # PIPA527701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 211 of Human BAG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 2; BAG-2; BAG-2 dJ417I1.2 BAG family molecular chaperone regulator 2 BAG-family molecular chaperone regulator-2 bcl-2-associated athanogene 2 dJ417I1.2 (BAG-family molecular chaperone regulator 2); BAG-family molecular chaperone regulator-2; Bcl-2-associated athanogene 2; BCL2-associated athanogene 2; dJ417I1.2 (BAG-family molecular chaperone regulator 2)
Gene Aliases: BAG-2; BAG2; dJ417I1.2
UniProt ID: (Human) O95816
Entrez Gene ID: (Human) 9532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.