Invitrogen
Manufacturer Code:PA5110832
Catalog # PIPA5110832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human BAG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 1; BAG family molecular chaperone regulator 1 BAG1 BAG-1 Bcl-2-associated athanogene 1 HAP RAP46 RP11-326F20.2; BAG-1; Bcl-2 associating athanogene-1 protein; Bcl-2-associated athanogene 1; Bcl-2-binding protein; BCL2-associated athanogene; glucocortoid receptor-associated protein RAP46; receptor-associated protein, 46-KD
Gene Aliases: BAG-1; BAG1; HAP; RAP46
UniProt ID: (Human) Q99933
Entrez Gene ID: (Human) 573
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.