Invitrogen
Manufacturer Code:PA529436
Catalog # PIPA529436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 105 and 441 of Human SMARCD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa BRG-1/Brm-associated factor subunit C; BAF60C; BAF60C CRACD3 Rsc6p 60 kDa BRG-1/Brm-associated factor subunit C BRG1-associated factor 60C SWI/SNF complex 60 kDa subunit C SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3 Swp73-like protein chromatin remodeling complex BAF60C s; BRG1-associated factor 60C; chromatin remodeling complex BAF60C subunit; mammalian chromatin remodeling complex BRG1-associated factor 60C; mBAF60c; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein
Gene Aliases: 1500001J14Rik; 2210409C08Rik; BAF60C; CRACD3; Rsc6p; SMARCD3
UniProt ID: (Human) Q6STE5, (Mouse) Q6P9Z1
Entrez Gene ID: (Human) 6604, (Mouse) 66993
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.