Invitrogen
Manufacturer Code:PA583758
Catalog # PIPA583758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACTL6B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor B; Actin-like protein 6B; Actin-related protein Baf53b; ACTL6 BAF53B; ArpNalpha; BAF53B; BRG1-associated factor 53B; hArpN alpha
Gene Aliases: ACTL6; ACTL6B; arpNalpha; BAF53B
UniProt ID: (Human) O94805
Entrez Gene ID: (Human) 51412
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.