Proteintech
Manufacturer Code:272151AP150UL
Catalog # 50-173-5606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BAF53B Fusion Protein Ag25745 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor B; actin like 6B Actin like protein 6B Actin related protein Baf53b ACTL6 ACTL6B ArpNalpha BAF53B BRG1 associated factor 53B; actin-like 6; Actin-like protein 6B; actin-like protein 6B {ECO:0000250|UniProtKB:O94805}; Actin-related protein Baf53b; actl6b {ECO:0000250|UniProtKB:O94805}; ArpN-alpha; ArpNa; ArpNalpha; BAF53B; BRG1-associated factor 53B; hArpN alpha
Gene Aliases: ACTL6; ACTL6B; ArpNa; arpNalpha; BAF53B
UniProt ID: (Human) O94805, (Rat) P86173, (Mouse) Q99MR0
Entrez Gene ID: (Human) 51412, (Rat) 288563, (Mouse) 83766
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.