Invitrogen
Manufacturer Code:MA518344
Catalog # PIMA518344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C4 |
Immunogen | ACTL6A (NP&lowbar817126 1 a.a. &bsim 80 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor A; actin-like 6A; Actin-like protein 6A; actin-related protein 4; Actin-related protein Baf53a; ACTL6A ACTL6B BAF53A BAF53B ACTL6 BAF53a BAF53b actin-like 6A actin-like 6B hArpN alpha actin-related protein ACTL6A ACTL6B 53 kDa BRG1-associated factor B ARPN-BETA MGC5382 Arp4 INO80K BAF complex 53 kDa subunit BAF53 BRG1-associated factor INO80 complex subunit K actin-related protein 4 hArpN beta; ArpNbeta; BAF complex 53 kDa subunit; BAF53; BAF53A; BRG1-associated factor 53A; hArpN beta; INO80 complex subunit K
Gene Aliases: ACTL6; ACTL6A; Arp4; ARPN-BETA; BAF53; BAF53A; INO80K
UniProt ID: (Human) O96019
Entrez Gene ID: (Human) 86
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.