Invitrogen
Manufacturer Code:PA586371
Catalog # PIPA586371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 211-260 of Human BAF53. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor A; actin-like 6; actin-like 6A; Actin-like protein 6A; Actin-like protein 6A 53 kDa BRG1-associated factor A Actin-related protein Baf53a ArpNbeta BRG1-associated factor 53A BAF53A INO80 complex subunit K ACTL6A BAF53 BAF53A INO80K; actin-related protein 4; Actin-related protein Baf53a; ArpNbeta; BAF complex 53 kDa subunit; BAF53; BAF53A; BRG1-associated factor 53A; BRG1/brm-associated factor 53A; hArpN beta; INO80 complex subunit K
Gene Aliases: 2810432C06Rik; ACTL6; ACTL6A; AI851094; ARP4; ARPN-BETA; BAF53; BAF53A; C79802; INO80K; RGD1307747
UniProt ID: (Human) O96019, (Mouse) Q9Z2N8
Entrez Gene ID: (Human) 86, (Mouse) 56456, (Rat) 361925
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.