Invitrogen
Manufacturer Code:PA519421
Catalog # PIPA519421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Human BAF53A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor A; actin-like 6; actin-like 6A; Actin-like protein 6A; actin-related protein 4; Actin-related protein Baf53a; ACTL6 ARPN-BETA Arp4 BAF53A INO80K MGC5382 53 kDa BRG1-associated factor A BAF complex 53 kDa subunit BAF53 BRG1-associated factor 53A INO80 complex subunit K actin-like protein 6A actin-related protein 4 actin-related protein Baf53a arpNbeta hArpN beta; ArpNbeta; BAF complex 53 kDa subunit; BAF53; BAF53A; BRG1-associated factor 53A; BRG1/brm-associated factor 53A; hArpN beta; INO80 complex subunit K
Gene Aliases: 2810432C06Rik; ACTL6; ACTL6A; AI851094; ARP4; ARPN-BETA; BAF53; BAF53A; C79802; INO80K; RGD1307747
UniProt ID: (Human) O96019, (Mouse) Q9Z2N8
Entrez Gene ID: (Human) 86, (Mouse) 56456, (Rat) 361925
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.