Novus Biologicals
Manufacturer Code:NBP299367DL594
Catalog # NB085009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human BAF53A. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53 kDa BRG1-associated factor A; actin-like 6A; actin-like 6A actin-like protein 6A actin-related protein 453 kDa BRG1-associated factor A Actin-related protein Baf53a Actl6 Arp4 ArpNbeta BAF complex 53 kDa subunit Baf53a BAF53AACTL6 BAF53MGC5382 BRG1-associated factor 53A hArpN beta INO80 complex subunit K INO80KARPN-BETA; Actin-like protein 6A; actin-related protein 4; Actin-related protein Baf53a; ArpNbeta; BAF complex 53 kDa subunit; BAF53; BAF53A; BRG1-associated factor 53A; hArpN beta; INO80 complex subunit K
Gene Aliases: ACTL6; ACTL6A; Arp4; ARPN-BETA; BAF53; BAF53A; INO80K
UniProt ID: (Human) O96019
Entrez Gene ID: (Human) 86
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.