Invitrogen
Manufacturer Code:PA541624
Catalog # PIPA541624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BACH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; Transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2)
Gene Aliases: BACH2; BTBD25; E030004N02Rik
UniProt ID: (Human) Q9BYV9, (Mouse) P97303
Entrez Gene ID: (Horse) 100065814, (Human) 60468, (Rabbit) 100351257, (Mouse) 12014, (Rat) 313125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.