Invitrogen
Manufacturer Code:PA523642
Catalog # PIPA523642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 815-841 amino acids from the C-terminal region of human BACH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB and CNC homolog 2; BTB and CNC homolog 2 transcription regulator protein BACH2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; Transcription regulator protein BACH2
Gene Aliases: BACH2; BTBD25
UniProt ID: (Human) Q9BYV9
Entrez Gene ID: (Human) 60468
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.