Invitrogen
Manufacturer Code:PA5100792
Catalog # PIPA5100792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BACH2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bach2 BTB and CNC homolog 2 BTB and CNC homology 1 basic leucine zipper transcription factor 2 BTB and CNC homology 2 BTB domain and CNC homolog 2 BTBD25 E030004N02Rik RGD1562865 similar to BTB and CNC homology 1 basic leucine zipper transcription factor 2 transcription regulator protein BACH2 Transcription regulator protein BACH2 (BTB and CNC homolog 2); BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; Transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2)
Gene Aliases: BACH2; BTBD25; E030004N02Rik
UniProt ID: (Human) Q9BYV9, (Mouse) P97303
Entrez Gene ID: (Human) 60468, (Mouse) 12014, (Rat) 313125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.