Novus Biologicals
Manufacturer Code:NBP270240AF594
Catalog # NP270240A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E11 |
Immunogen | Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: basic region leucine zipper transcriptional regulator BACH1; basic region leucine zipper transcriptional regulator BACH1 BTB and CNC homolog 1 BTB and CNC homology 1 basic leucine zipper transcription factor 1 HA2303 transcription regulator protein BACH1 transcription regulator protein10BACH-1; BTB and CNC homolog 1; BTB and CNC homology 1, basic leucine zipper transcription factor 1; HA2303; Transcription regulator protein BACH1
Gene Aliases: BACH-1; BACH1; BTBD24
UniProt ID: (Human) O14867
Entrez Gene ID: (Human) 571
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.