Abnova
Manufacturer Code:H00025825A01
Catalog # 89-003-138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | BACE2 (NP_036237 306 a.a. ~ 395 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant BACE2.
For Research Use Only
Protein Aliases: 56 kDa aspartic-like protease; AEPLC; ALP56; ASP1; ASP21; Aspartic-like protease 56 kDa; Aspartyl protease 1; BAE2; Beta-secretase 2; beta-site amyloid beta A4 precursor protein-cleaving enzyme 2; Beta-site amyloid precursor protein cleaving enzyme 2; Beta-site APP cleaving enzyme 2; CDA13; CEAP1; Down region aspartic protease; Down syndrome region aspartic protease; DRAP; memapsin 1; Memapsin-1; Membrane-associated aspartic protease 1; Theta-secretase; transmembrane aspartic proteinase Asp1
Gene Aliases: AEPLC; ALP56; ASP1; ASP21; BACE2; BAE2; CDA13; CEAP1; DRAP; UNQ418/PRO852
UniProt ID: (Human) Q9Y5Z0
Entrez Gene ID: (Human) 25825
Molecular Function: aspartic protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.