Invitrogen
Manufacturer Code:MA1177
Catalog # PIMA1177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C1C3 |
Immunogen | Purified recombit fragment of Human BACE1 (AA: 112-324) expressed in E. Coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APP beta-secretase; asp 2; ASP2; Aspartyl protease 2; BACE ASP2 HSPC104 C76936; Beta-secretase 1; beta-secretase 1 precursor variant 1; beta-site amyloid beta A4 precursor protein-cleaving enzyme; Beta-site amyloid precursor protein cleaving enzyme 1; Beta-site APP cleaving enzyme 1; beta-site APP-cleaving enzyme 1; Memapsin-2; Membrane-associated aspartic protease 2; protease; transmembrane aspartic proteinase Asp2
Gene Aliases: ASP2; BACE; BACE1; C76936; HSPC104; KIAA1149
UniProt ID: (Human) P56817, (Rat) P56819, (Mouse) P56818
Entrez Gene ID: (Human) 23621, (Dog) 489390, (Rat) 29392, (Mouse) 23821
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.