R&D Systems
Manufacturer Code:MAB931SP
Catalog # MAB931SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 137612 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human BACE-1 Thr22-Tyr460 Accession # P56817 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APP beta-secretase; APP beta-secretase ASP2 Aspartyl protease 2 BACE1 BACEAsp 2 beta-secretase 1 beta-secretase 1 precursor variant 1 beta-site amyloid beta A4 precursor protein-cleaving enzyme Beta-site amyloid precursor protein cleaving enzyme 1 Beta-site APP cleaving enzyme 1 beta-site APP-cleaving enzyme beta-site APP-cleaving enzyme 1 EC 3.4.23 EC 3.4.23.46 FLJ90568 HSPC104 KIAA1149 Memapsin-2 Membrane-associated aspartic protease 2 transmembrane aspartic proteinase Asp2; asp 2; ASP2; Aspartyl protease 2; Beta-secretase 1; beta-secretase 1 precursor variant 1; beta-site amyloid beta A4 precursor protein-cleaving enzyme; Beta-site amyloid precursor protein cleaving enzyme 1; Beta-site APP cleaving enzyme 1; beta-site APP-cleaving enzyme 1; Memapsin-2; Membrane-associated aspartic protease 2; transmembrane aspartic proteinase Asp2
Gene Aliases: ASP2; BACE; BACE1; HSPC104; KIAA1149
UniProt ID: (Human) P56817
Entrez Gene ID: (Human) 23621
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.