Invitrogen
Manufacturer Code:PA585503
Catalog # PIPA585503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human HSPC142. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRCA1-A complex subunit MERIT40; BRISC and BRCA1-A complex member 1; C19orf62 antibody FLJ20571 antibody MERIT40 antibody NBA1 antibody BABAM1 antibody new component of the BRCA1-A complex antibody BRISC and BRCA1-A complex member 1 antibody new component of the BRCAA1 A complex antibody BRCA1-A complex subunit MERIT40 antibody mediator of Rap80 interactions and targeting 40 kDa antibody new component of the BRCA1 A complex antibody mediator of RAP80 interactions and targeting subunit of 40 kDa antibody BRISC and BRCA1 A complex member 1 antibody; mediator of Rap80 interactions and targeting 40 kDa; Mediator of RAP80 interactions and targeting subunit of 40 kDa; new component of the BRCA1 A complex; New component of the BRCA1-A complex; new component of the BRCAA1 A complex
Gene Aliases: BABAM1; C19orf62; HSPC142; MERIT40; NBA1
UniProt ID: (Human) Q9NWV8
Entrez Gene ID: (Human) 29086
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.