Invitrogen
Manufacturer Code:PA5105264
Catalog # PIPA5105264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MERIT40 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5430437P03Rik BABAM1 BRCA1-A complex subunit MERIT40 BRISC and BRCA1 A complex member 1 BRISC and BRCA1-A complex member 1 C19orf62 HSPC142 mediator of Rap80 interactions and targeting 40 kDa mediator of RAP80 interactions and targeting subunit of 40 kDa MERIT40 NBA1 new component of the BRCA1 A complex new component of the BRCA1-A complex new component of the BRCAA1 A complex; BRCA1-A complex subunit MERIT40; BRISC and BRCA1-A complex member 1; mediator of Rap80 interactions and targeting 40 kDa; Mediator of RAP80 interactions and targeting subunit of 40 kDa; new component of the BRCA1 A complex; New component of the BRCA1-A complex; new component of the BRCAA1 A complex
Gene Aliases: 5430437P03Rik; BABAM1; C19orf62; HSPC142; MERIT40; NBA1
UniProt ID: (Human) Q9NWV8, (Mouse) Q3UI43, (Rat) Q5XIJ6
Entrez Gene ID: (Human) 29086, (Mouse) 68251, (Rat) 290631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.