Invitrogen
Manufacturer Code:PA5103658
Catalog # PIPA5103658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BAAT corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI118337 AI158864 Baat BACAT BAT bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase) bile acid CoA:amino acid N-acyltransferase bile acid Coenzyme A: amino acid N-acyltransferase (glycine N-choloyltransferase) Bile acid-CoA thioesterase Bile acid-CoA:amino acid N-acyltransferase bile acid-Coenzyme A dehydrogenase: amino acid n-acyltransferase bile acid-Coenzyme A: amino acid N-acyltransferase Choloyl-CoA hydrolase Glycine N-choloyltransferase Kan-1 Long-chain fatty-acyl-CoA hydrolase taurine N-acyltransferase; BACAT; bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase); bile acid CoA:amino acid N-acyltransferase; bile acid Coenzyme A: amino acid N-acyltransferase (glycine N-choloyltransferase); Bile acid-CoA thioesterase; Bile acid-CoA:amino acid N-acyltransferase; bile acid-Coenzyme A dehydrogenase: amino acid n-acyltransferase; Choloyl-CoA hydrolase; Glycine N-choloyltransferase; Long-chain fatty-acyl-CoA hydrolase; taurine N-acyltransferase
Gene Aliases: AI118337; AI158864; BAAT; BACAT; BAT
UniProt ID: (Human) Q14032, (Mouse) O08833
Entrez Gene ID: (Human) 570, (Mouse) 12012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.