Abnova
Manufacturer Code:H00000570A01
Catalog # 89-104-136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 258 a.a. to 355 a.a) partial recombinant protein with GST tag.,BAAT (NP_001692 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant BAAT.
For Research Use Only
Protein Aliases: BACAT; BAT; bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase); bile acid CoA:amino acid N-acyltransferase; bile acid Coenzyme A: amino acid N-acyltransferase (glycine N-choloyltransferase); Bile acid-CoA thioesterase; Bile acid-CoA:amino acid N-acyltransferase; Choloyl-CoA hydrolase; FLJ20300; Glycine N-choloyltransferase; Long-chain fatty-acyl-CoA hydrolase; MGC104432
Gene Aliases: BAAT; BACAT; BAT
UniProt ID: (Human) Q14032
Entrez Gene ID: (Human) 570
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.