Invitrogen
Manufacturer Code:PA518435
Catalog # PIPA518435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KQNGLQTTEAKRDAK) corresponding to the internal amino acids of BAALC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BAALC brain and acute leukemia cytoplasmic HGNC:14333 FLJ12015; Brain and acute leukemia cytoplasmic protein
Gene Aliases: 2810457D07Rik; BAALC
UniProt ID: (Human) Q8WXS3, (Mouse) Q8VHV1
Entrez Gene ID: (Human) 79870, (Mouse) 118452
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.