Invitrogen
Manufacturer Code:PA576494
Catalog # PIPA576494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human B9D1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B9 domain-containing protein 1; B9 domain-containing protein 1 MKS1-related protein 1 B9D1 MKSR1; B9 protein domain 1; Endothelial precursor cells protein B9; endothelial precursor protein B9; MKS1-related protein 1
Gene Aliases: AW045994; B9; B9D1; EPPB9; MKS9; MKSR1
UniProt ID: (Human) Q9UPM9, (Rat) P0C5J2, (Mouse) Q9R1S0
Entrez Gene ID: (Human) 27077, (Rat) 287383, (Mouse) 27078
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.