Invitrogen
Manufacturer Code:PA554645
Catalog # PIPA554645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human B9D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B9 domain-containing protein 1; B9 protein domain 1; endothelial precursor protein B9; MKS1-related protein 1
Gene Aliases: B9; B9D1; EPPB9; MKS9; MKSR1
UniProt ID: (Human) Q9UPM9
Entrez Gene ID: (Human) 27077
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.