Novus Biologicals
Manufacturer Code:NBP29282120UL
Catalog # NB027509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human B9D1 (NP_056496.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B9 B9 domain-containing protein 1 B9 protein domain 1 endothelial precursor protein B9 EPPB9 MKS1-related protein 1 MKSR1; B9 domain-containing protein 1; B9 protein domain 1; endothelial precursor protein B9; MKS1-related protein 1
Gene Aliases: B9; B9D1; EPPB9; MKS9; MKSR1
UniProt ID: (Human) Q9UPM9
Entrez Gene ID: (Human) 27077
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.