Invitrogen
Manufacturer Code:16594982
Catalog # 50-146-45
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Control (Ctrl) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H74 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B7 family member, H4; B7 homolog 4; B7 superfamily member 1; B7-H4; B7-H4 B7H4 B7S1 B7X B7h.5 FLJ22418 PRO1291 RP11-229A19.4 VCTN1; B7h.5; Immune costimulatory protein B7-H4; Protein B7S1; T cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; V-set domain-containing T-cell activation inhibitor 1; VTCN1
Gene Aliases: B7-H4; B7h.5; B7H4; B7S1; B7X; PRO1291; UNQ659/PRO1291; VCTN1; VTCN1
UniProt ID: (Human) Q7Z7D3
Entrez Gene ID: (Human) 79679
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.