R&D Systems
Manufacturer Code:FAB6576P100
Catalog # FAB6576P100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 935317 |
Immunogen | NS0 mouse myeloma cell line transfected with human B7-H4 Phe29-Ser258 Accession # Q7Z7D3 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B7 family member, H4; B7 homolog 4; B7 superfamily member 1; B7-H4; B7h.5; B7h.5 B7H4 B7-H4 B7H4T-cell costimulatory molecule B7x B7S1 B7S1VCTN1 B7x B7XPRO1291 FLJ22418 Immune costimulatory protein B7-H4 Protein B7S1 T cell costimulatory molecule B7x V-set domain containing T cell activation inhibitor 1 V-set domain-containing T-cell activation inhibitor 1 Vtcn1; Immune costimulatory protein B7-H4; Protein B7S1; T cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; V-set domain-containing T-cell activation inhibitor 1; VTCN1
Gene Aliases: B7-H4; B7h.5; B7H4; B7S1; B7X; PRO1291; UNQ659/PRO1291; VCTN1; VTCN1
UniProt ID: (Human) Q7Z7D3
Entrez Gene ID: (Human) 79679
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.