R&D Systems
Manufacturer Code:FAB741G100UG
Catalog # FAB741G10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | GL1 |
Immunogen | LPS-activated mouse B cells |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rat Monoclonal Antibody
Protein Aliases: Activation B7-2 antigen; Activation B7-2 antigen B70 B72 B7-2 B7-2 antigen B-lymphocyte activation antigen B7-2 BU63 CD28 antigen ligand 2 CD28LG2B7-2 antigen) CD86 CD86 antigen CD86 molecule CTLA-4 counter-receptor B7.2 FUN-1 LAB72 MGC34413 T-lymphocyte activation antigen CD86; B-lymphocyte activation antigen B7-2; B70; BU63; CD86; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CTLA-4 counter-receptor B7.2; FUN-1; T-lymphocyte activation antigen CD86
Gene Aliases: B7-2; B7.2; B70; CD28LG2; CD86; LAB72
UniProt ID: (Human) P42081
Entrez Gene ID: (Human) 942
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.