R&D Systems
Manufacturer Code:AF740SP
Catalog # AF740SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse B7-1/CD80 Asp37-Lys245 Accession # Q00609 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Activation B7-1 antigen; Activation B7-1 antigen B7 B71 BB1 CD28LG1 CD28LGB7-1 antigen) CD80 CD80 antigen CD80 molecule costimulatory factor CD80 costimulatory molecule variant IgV-CD80 CTLA-4 counter-receptor B7.1 T-lymphocyte activation antigen CD80; B-lymphocyte activation antigen B7; B7; BB1; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; T-lymphocyte activation antigen CD80
Gene Aliases: B7; B7-1; B7.1; BB1; CD28LG; CD28LG1; CD80; LAB7
UniProt ID: (Human) P33681
Entrez Gene ID: (Human) 941
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.