Invitrogen
Manufacturer Code:PA575761
Catalog # PIPA575761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human B4GALT6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b4Gal-T6; beta-1, 4-galactosyltransferase VI; Beta-1,4-galactosyltransferase 6; Beta-1,4-GalTase 6; Beta-14-galactosyltransferase 6 Beta-14-GalTase 6 Beta4Gal-T6 b4Gal-T6 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 6 UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 6 Including the following 1 domains:Glucosylceramide beta-14-galactosyltransferase Lactosylceramide synthase LacCer synthase UDP-Gal:glucosylceramide beta-14-galactosyltransferase B4GALT6; beta4Gal-T6; beta4GalT-VI; Glucosylceramide beta-1,4-galactosyltransferase; LacCer synthase; Lactosylceramide synthase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 6; UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6
Gene Aliases: AA536803; AU022389; B4Gal-T6; B4GALT6; beta4Gal-T6
UniProt ID: (Human) Q9UBX8, (Mouse) Q9WVK5
Entrez Gene ID: (Human) 9331, (Mouse) 56386
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.