Novus Biologicals
Manufacturer Code:NBP299252B
Catalog # NB059439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of mouse B4GALT6. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B4Gal-T6 beta-14-galactosyltransferase 6 Beta-14-GalTase 6 Beta4Gal-T6 beta4GalT-VI EC 2.4.1.- UDP-Gal:betaGlcNAc beta 14- galactosyltransferase polypeptide 6 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 6 UDP-Gal:glucosylceramide beta-14-galactosyltransferase UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 6; Beta-1,4-galactosyltransferase 6; Beta-1,4-GalTase 6; beta4GalT-VI; Glucosylceramide beta-1,4-galactosyltransferase; LacCer synthase; Lactosylceramide synthase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 6; UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6
Gene Aliases: B4Gal-T6; B4GALT6; beta4Gal-T6
UniProt ID: (Human) Q9UBX8
Entrez Gene ID: (Human) 9331
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.