Novus Biologicals
Manufacturer Code:NBP271385FR
Catalog # NBP271385FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8B6 |
Immunogen | Recombinant protein fragment corresponding to amino acids 40-322 of human B4GALT4 (NP_003769) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: B4Gal-T4 beta-14-galactosyltransferase 4 Beta-14-GalTase 4 beta4Gal-T4 beta-N-acetylglucosaminyl-glycolipid beta-14-galactosyltransferase 4 EC 2.4.1.- UDP-Gal:betaGlcNAc beta 14- galactosyltransferase 4 UDP-Gal:betaGlcNAc beta 14- galactosyltransferase polypeptide 4 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 4 UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 4; Beta-1,4-galactosyltransferase 4; Beta-1,4-GalTase 4; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4; Lactotriaosylceramide beta-1,4-galactosyltransferase; N-acetyllactosamine synthase; Nal synthase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4
Gene Aliases: B4Gal-T4; B4GALT4; beta4Gal-T4; UNQ552/PRO1109
UniProt ID: (Human) O60513
Entrez Gene ID: (Human) 8702
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.