Invitrogen
Manufacturer Code:PA568112
Catalog # PIPA568112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human B4GALT4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b4Gal-T4; B4Gal-T4 beta4Gal-T4 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 4 UDP-Gal:betaGlcNAc beta 14- galactosyltransferase 4 UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 4 beta-14-GalTase 4 beta-14-galactosyltransferase 4 beta-N-acetylglucosaminyl; Beta-1,4-galactosyltransferase 4; beta-1,4-galactosyltransferase IV; Beta-1,4-GalTase 4; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4; beta4Gal-T4; Lactotriaosylceramide beta-1,4-galactosyltransferase; N-acetyllactosamine synthase; Nal synthase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4
Gene Aliases: 9130402O08Rik; B4Gal-T4; B4galt-IV; B4GALT4; beta4Gal-T4; beta4GalT-IV; UNQ552/PRO1109
UniProt ID: (Human) O60513, (Mouse) Q9JJ04, (Rat) Q66HH1
Entrez Gene ID: (Human) 8702, (Mouse) 56375, (Rat) 303923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.