Novus Biologicals
Manufacturer Code:NBP271386AF700
Catalog # N271386A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G9 |
Immunogen | Recombinant protein fragment corresponding to amino acids 100-393 of human B4GALT3 (NP_003770) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: b4Gal-T3; b4Gal-T3 beta-14-galactosyltransferase 3 Beta-14-GalTase 3 Beta4Gal-T3 beta-N-acetylglucosaminyl-glycolipid beta-14-galactosyltransferase 3 EC 2.4.1.- UDP-Gal:betaGlcNAc beta 14- galactosyltransferase polypeptide 3 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 3 UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 3; Beta-1,4-galactosyltransferase 3; Beta-1,4-GalTase 3; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; N-acetyllactosamine synthase; Nal synthase; Neolactotriaosylceramide beta-1,4-galactosyltransferase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3
Gene Aliases: B4GALT3; beta4Gal-T3
UniProt ID: (Human) O60512
Entrez Gene ID: (Human) 8703
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.