Proteintech
Manufacturer Code:110411AP150UL
Catalog # 50-172-6822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | B4GALT3 Fusion Protein Ag1487 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b4Gal T3 B4GALT3 Beta 14 GalTase 3 beta4Gal T3; b4Gal-T3; Beta-1,4-galactosyltransferase 3; beta-1,4-galactosyltransferase III; Beta-1,4-GalTase 3; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; beta4Gal-T3; beta4GalT-III; N-acetyllactosamine synthase; Nal synthase; Neolactotriaosylceramide beta-1,4-galactosyltransferase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 3; beta-1,4-galactosyltransferase III; beta4GalT-III; expressed sequence tag mouse EST 6; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3
Gene Aliases: 9530061M23Rik; AA104562; AW125175; B4GALT3; beta4Gal-T3; ESTM26; ESTM6; R74981
UniProt ID: (Human) O60512, (Rat) Q6P768, (Mouse) Q91YY2
Entrez Gene ID: (Human) 8703, (Rat) 494342, (Mouse) 57370
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.