Abnova
Manufacturer Code:H00008703K
Catalog # 89-203-549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human B4GALT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human B4GALT3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: b4Gal-T3; Beta-1,4-galactosyltransferase 3; Beta-1,4-GalTase 3; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; beta4Gal-T3; N-acetyllactosamine synthase; Nal synthase; Neolactotriaosylceramide beta-1,4-galactosyltransferase; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3
Gene Aliases: B4GALT3; beta4Gal-T3
UniProt ID: (Human) O60512
Entrez Gene ID: (Human) 8703
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.