Invitrogen
Manufacturer Code:PA5117336
Catalog # PIPA5117336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human B4GALT1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b14-Galactosyltransferase I B-14-GalT B-14-GalT1 B4galt1 b4Gal-T1 beta-14-galactosyltransferase 1 beta-14-GalT beta-14-GalT1 Beta-14-GalTase 1 beta4Gal-T1 Beta-N-acetylglucosaminyl-glycolipid beta-14-galactosyltransferase Beta-N-acetylglucosaminylglycopeptide beta-14-galactosyltransferase CDG2D galactosyltransferase 2 beta 1 4 GalT Ggtb GGTB2 glycoprotein galactosyltransferase beta 1 4 glycoprotein-4-beta-galactosyltransferase 2 Golgi complex GT1 GTB lactose synthase Lactose synthase A protein N-acetyllactosamine synthase Nal synthase Processed beta-1 Processed beta-14-galactosyltransferase 1 UDP-Gal:betaGlcNAc beta 14- galactosyltransferase polypeptide 1 UDP-Gal:beta-GlcNAc beta-14-galactosyltransferase 1 UDP-galactose:beta-N-acetylglucosamine beta-14-galactosyltransferase 1; Beta-1,4-galactosyltransferase 1; Beta-1,4-GalTase 1; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; glycoprotein-4-beta-galactosyltransferase 2; lactose synthase; Lactose synthase A protein; N-acetyllactosamine synthase; Nal synthase; Neolactotriaosylceramide beta-1,4-galactosyltransferase; Processed beta-1,4-galactosyltransferase 1; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1
Gene Aliases: B4GAL-T1; B4GALT1; beta4Gal-T1; CDG2D; GGTB2; GT1; GTB
UniProt ID: (Human) P15291
Entrez Gene ID: (Human) 2683
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.