Invitrogen
Manufacturer Code:PA570775
Catalog # PIPA570775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human B4GALNT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; Beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-4-N-acetylgalactosaminyltransferase; GALGT2; Sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase
Gene Aliases: B4GALNT2; B4GALT; GALGT2
UniProt ID: (Human) Q8NHY0
Entrez Gene ID: (Human) 124872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.